missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC26A6 Full-length ORF (NP_599025.2, 1 a.a. - 758 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_599025.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 65010 |
Molecular Weight (g/mol) | 109.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16113863
|
Abnova™
H00065010-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16103863
|
Abnova™
H00065010-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene belongs to the solute carrier 26 family, whose members encode anion transporter proteins. This particular family member encodes a protein involved in transporting chloride, oxalate, sulfate and bicarbonate. Several alternatively spliced transcript variants of this gene, encoding distinct isoforms, have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Sequence: MGLADASGPRDTQALLSATQAMDLRRRDYHMERPLLNQEHLEELGRWGSAPRTHQWRTWLQCSRARAYALLLQHLPVLVWLPRYPVRDWLLGDLLSGLSVAIMQLPQGLAYALLAGLPPVFGLYSSFYPVFIYFLFGTSRHISVGTFAVMSVMVGSVTESLAPQALNDSMINETARDAARVQVASTLSVLVGLFQVGLGLIHFGFVVTYLSEPLVRGYTTAAAVQVFVSQLKYVFGLHLSSHSGPLSLIYTVLEVCWKLPQSKVGTVVTAAVAGVVLVVVKLLNDKLQQQLPMPIPGELLTLIGATGISYGMGLKHRFEVDVVGNIPAGLVPPVAPNTQLFSKLVGSAFTIAVVGFAIAISLGKIFALRHGYRVDSNQELVALGLSNLIGGIFQCFPVSCSMSRSLVQESTGGNSQVAGAISSLFILLIIVKLGELFHDLPKAVLAAIIIVNLKGMLRQLSDMRSLWKANRADLLIWLVTFTATILLNLDLGLVVAVIFSLLLVVVRTQMPHYSVLGQVPDTDIYRDVAEYSEAKEVRGVKVFRSSATVYFANAEFYSDALKQRCGVDVDFLISQKKKLLKKQEQLKLKQLQKEEKLRKQAASPKGASVSINVNTSLEDMRSNNVEDCKMMVSSGDKMEDATANGQEDSKAPDGSTLKALGLPQPDFHSLILDLGALSFVDTVCLKSLKNIFHDFREIEVEVYMAACHSPVVSQLEAGHFFDASITKKHLFASVHDAVTFALQHPRPVPDSPVSVTRLSpecifications
NP_599025.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
109.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGLADASGPRDTQALLSATQAMDLRRRDYHMERPLLNQEHLEELGRWGSAPRTHQWRTWLQCSRARAYALLLQHLPVLVWLPRYPVRDWLLGDLLSGLSVAIMQLPQGLAYALLAGLPPVFGLYSSFYPVFIYFLFGTSRHISVGTFAVMSVMVGSVTESLAPQALNDSMINETARDAARVQVASTLSVLVGLFQVGLGLIHFGFVVTYLSEPLVRGYTTAAAVQVFVSQLKYVFGLHLSSHSGPLSLIYTVLEVCWKLPQSKVGTVVTAAVAGVVLVVVKLLNDKLQQQLPMPIPGELLTLIGATGISYGMGLKHRFEVDVVGNIPAGLVPPVAPNTQLFSKLVGSAFTIAVVGFAIAISLGKIFALRHGYRVDSNQELVALGLSNLIGGIFQCFPVSCSMSRSLVQESTGGNSQVAGAISSLFILLIIVKLGELFHDLPKAVLAAIIIVNLKGMLRQLSDMRSLWKANRADLLIWLVTFTATILLNLDLGLVVAVIFSLLLVVVRTQMPHYSVLGQVPDTDIYRDVAEYSEAKEVRGVKVFRSSATVYFANAEFYSDALKQRCGVDVDFLISQKKKLLKKQEQLKLKQLQKEEKLRKQAASPKGASVSINVNTSLEDMRSNNVEDCKMMVSSGDKMEDATANGQEDSKAPDGSTLKALGLPQPDFHSLILDLGALSFVDTVCLKSLKNIFHDFREIEVEVYMAACHSPVVSQLEAGHFFDASITKKHLFASVHDAVTFALQHPRPVPDSPVSVTRL | |
RUO | |
SLC26A6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
65010 | |
SLC26A6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586E1422 | |
SLC26A6 | |
Recombinant | |
wheat germ expression system |