missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC12A1 Full-length ORF (ENSP00000331550, 1 a.a. - 430 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006557-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190802

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

The sodium-potassium-chloride cotransporter isoform 2 is kidney-specific and is found on the apical membrane of the thick ascending limb of Henle's loop and the macula densa. It accounts for most of the NaCl resorption with the stoichiometry of 1Na:1K:2Cl and is sensitive to such diuretics as furosemide and bumetanide. Some Bartter-like syndromes result from defects in this gene. [provided by RefSeq]

Sequence: MSLNNSSNVFLDSVPSNTNRFQVSVINENHESSAAADDNTDPPHYEETSFGDEAQKRLRISFRPGNQECYDNFLQSGETAKTDASFHAYDSHTNTYYLQTFGHNTMDAVPKIEYYRNTGSISGPKVNRPSLLEIHEQLAKNVAVTPSSADRVANGDGIPGDEQAENKEDDQAGVVKFGWVKGVLVRCMLNIWGVMLFIRLSWIVGEAGIGLGVLIILLSTMVTSITGLSTSAIATNGFVRGGGAYYLISRSLGPEFGGSIGLIFAFANAVAVAMYVVGFAETVVDLLKESDSMMVDPTNDIRIIGSITVVILLGISVAGMEWEAKAQVILLVILLIAIANFFIGTVIPSNNEKKSRGFFNYQASIFAENFGPRFTKGEGFFSVFAIFFPAATGILAGANISGDLEALRKQGASPLPQGAQSRRRRQTSLI
Specifications

Specifications

ENSP00000331550
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
72.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BSC1/MGC48843/NKCC2
SLC12A1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
6557
SLC12A1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSLNNSSNVFLDSVPSNTNRFQVSVINENHESSAAADDNTDPPHYEETSFGDEAQKRLRISFRPGNQECYDNFLQSGETAKTDASFHAYDSHTNTYYLQTFGHNTMDAVPKIEYYRNTGSISGPKVNRPSLLEIHEQLAKNVAVTPSSADRVANGDGIPGDEQAENKEDDQAGVVKFGWVKGVLVRCMLNIWGVMLFIRLSWIVGEAGIGLGVLIILLSTMVTSITGLSTSAIATNGFVRGGGAYYLISRSLGPEFGGSIGLIFAFANAVAVAMYVVGFAETVVDLLKESDSMMVDPTNDIRIIGSITVVILLGISVAGMEWEAKAQVILLVILLIAIANFFIGTVIPSNNEKKSRGFFNYQASIFAENFGPRFTKGEGFFSVFAIFFPAATGILAGANISGDLEALRKQGASPLPQGAQSRRRRQTSLI
RUO
SLC12A1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers