missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Serum Albumin Native Protein

Product Code. 18001364 Shop All Bio Techne Products
Click to view available options
:
1mg; Texas Red
This item is not returnable. View return policy

Product Code. 18001364

Brand: Novus Biologicals™ NBP196791

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

Native Human Serum Albumin. The Human Serum Albumin Native Protein has been validated for the following applications: Western Blot, Fluorophore-linked immunosorbent assay, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02768
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10mg/mL Polyethylene Glycol (PEG-8000)
Gene ID (Entrez) 280717
Name Bovine Serum Albumin protein
Storage Requirements 4°C in the dark.
Cross Reactivity Human
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.