missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEMA3B Partial ORF (NP_004627, 651 a.a. - 748 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007869-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16158035

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

The semaphorin/collapsin family of molecules plays a critical role in the guidance of growth cones during neuronal development. The secreted protein encoded by this gene family member is important in axonal guidance and has been shown to act as a tumor suppressor by inducing apoptosis. [provided by RefSeq]

Sequence: FTQPLRRLSLHVLSATQAERLARAEEAAPAAPPGPKLWYRDFLQLVEPGGGGSANSLRMCRPQPALQSLPLESRRKGRNRRTHAPEPRAERGPRSATH
Specifications

Specifications

NP_004627
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FTQPLRRLSLHVLSATQAERLARAEEAAPAAPPGPKLWYRDFLQLVEPGGGGSANSLRMCRPQPALQSLPLESRRKGRNRRTHAPEPRAERGPRSATH
RUO
SEMA3B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7869
SEMA3B (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ34863/LUCA-1/SEMA5/SEMAA/SemA/semaV
SEMA3B
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers