missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SEMA3B Partial ORF (NP_004627, 651 a.a. - 748 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_004627 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7869 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16158035
|
Abnova™
H00007869-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16148035
|
Abnova™
H00007869-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The semaphorin/collapsin family of molecules plays a critical role in the guidance of growth cones during neuronal development. The secreted protein encoded by this gene family member is important in axonal guidance and has been shown to act as a tumor suppressor by inducing apoptosis. [provided by RefSeq]
Sequence: FTQPLRRLSLHVLSATQAERLARAEEAAPAAPPGPKLWYRDFLQLVEPGGGGSANSLRMCRPQPALQSLPLESRRKGRNRRTHAPEPRAERGPRSATHSpecifications
NP_004627 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ34863/LUCA-1/SEMA5/SEMAA/SemA/semaV | |
SEMA3B | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
7869 | |
SEMA3B (Human) Recombinant Protein (Q01) | |
FTQPLRRLSLHVLSATQAERLARAEEAAPAAPPGPKLWYRDFLQLVEPGGGGSANSLRMCRPQPALQSLPLESRRKGRNRRTHAPEPRAERGPRSATH | |
RUO | |
SEMA3B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |