missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RPL12 Partial ORF (NP_000967, 66 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006136-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16104555

  • 528.00€ / 25µg

Estimated Shipment: 18-07-2023
to see stock.
Add to basket

Description

Description

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L11P family of ribosomal proteins. It is located in the cytoplasm. The protein binds directly to the 26S rRNA. This gene is co-transcribed with the U65 snoRNA, which is located in its fourth intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq]

Sequence: NRQAQIEVVPSASALIIKALKEPPRDRKKQKNIKHSGNITFDEIVNIARQMRHRSLARELSGTIKEILGTAQSVGCNVDGRHPHDIIDDINSGAVECPAS
Specifications

Specifications

NP_000967
Liquid
6136
RPL12 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RPL12
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NRQAQIEVVPSASALIIKALKEPPRDRKKQKNIKHSGNITFDEIVNIARQMRHRSLARELSGTIKEILGTAQSVGCNVDGRHPHDIIDDINSGAVECPAS
RUO
RPL12
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers