Learn More
Abnova™ Human RNF14 Partial ORF (NP_004281, 217 a.a. - 316 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009604-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein interacts with androgen receptor (AR) and may function as a coactivator that induces AR target gene expression in prostate. A dominant negative mutant of this gene has been demonstrated to inhibit the AR-mediated growth of prostate cancer. This protein also interacts with class III ubiquitin-conjugating enzymes (E2s) and may act as a ubiquitin-ligase (E3) in the ubiquitination of certain nuclear proteins. Five alternatively spliced transcript variants encoding two distinct isoforms have been reported. [provided by RefSeq]
Sequence: LFLCSICFCEKLGSECMYFLECRHVYCKACLKDYFEIQIRDGQVQCLNCPEPKCPSVATPGQVKELVEAELFARYDRLLLQSSLDLMADVVYCPRPCCQLSpecifications
NP_004281 | |
Liquid | |
9604 | |
RNF14 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARA54/FLJ26004/HFB30/HRIHFB2038/TRIAD2 | |
RNF14 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LFLCSICFCEKLGSECMYFLECRHVYCKACLKDYFEIQIRDGQVQCLNCPEPKCPSVATPGQVKELVEAELFARYDRLLLQSSLDLMADVVYCPRPCCQL | |
RUO | |
RNF14 | |
Wheat Germ (in vitro) | |
GST |