missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNASEN Partial ORF (NP_037367.2, 1099 a.a. - 1196 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00029102-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16106016

  • 349.00€ / 10µg

Estimated Shipment: 18-07-2023
to see stock.
Add to basket

Description

Description

Members of the ribonuclease III superfamily of double-stranded (ds) RNA-specific endoribonucleases participate in diverse RNA maturation and decay pathways in eukaryotic and prokaryotic cells (Fortin et al., 2002 [PubMed 12191433]). The RNase III Drosha is the core nuclease that executes the initiation step of microRNA (miRNA) processing in the nucleus (Lee et al., 2003 [PubMed 14508493]).[supplied by OMIM]

Sequence: ETSPVLQKLTEFEEAIGVIFTHVRLLARAFTLRTVGFNHLTLGHNQRMEFLGDSIMQLVATEYLFIHFPDHHEGHLTLLRSSLVNNRTQAKVAEELGM
Specifications

Specifications

NP_037367.2
Liquid
29102
RNASEN (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DROSHA/ETOHI2/HSA242976/RANSE3L/RN3/RNASE3L
RNASEN
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ETSPVLQKLTEFEEAIGVIFTHVRLLARAFTLRTVGFNHLTLGHNQRMEFLGDSIMQLVATEYLFIHFPDHHEGHLTLLRSSLVNNRTQAKVAEELGM
RUO
RNASEN
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers