missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNASEN Partial ORF (NP_037367.2, 1099 a.a. - 1196 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

349.00€ - 528.00€

Specifications

Accession Number NP_037367.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 29102
Molecular Weight (g/mol) 36.52kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16116016
View Documents Promotion Details
Abnova™
H00029102-Q01.25UG
25 ug
528.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-11-2023
to see stock.
Add to basket
 
16106016
View Documents Promotion Details
Abnova™
H00029102-Q01.10UG
10 ug
349.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-11-2023
to see stock.
Add to basket
 
Description

Description

Members of the ribonuclease III superfamily of double-stranded (ds) RNA-specific endoribonucleases participate in diverse RNA maturation and decay pathways in eukaryotic and prokaryotic cells (Fortin et al., 2002 [PubMed 12191433]). The RNase III Drosha is the core nuclease that executes the initiation step of microRNA (miRNA) processing in the nucleus (Lee et al., 2003 [PubMed 14508493]).[supplied by OMIM]

Sequence: ETSPVLQKLTEFEEAIGVIFTHVRLLARAFTLRTVGFNHLTLGHNQRMEFLGDSIMQLVATEYLFIHFPDHHEGHLTLLRSSLVNNRTQAKVAEELGM
Specifications

Specifications

NP_037367.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DROSHA/ETOHI2/HSA242976/RANSE3L/RN3/RNASE3L
RNASEN
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
29102
RNASEN (Human) Recombinant Protein (Q01)
ETSPVLQKLTEFEEAIGVIFTHVRLLARAFTLRTVGFNHLTLGHNQRMEFLGDSIMQLVATEYLFIHFPDHHEGHLTLLRSSLVNNRTQAKVAEELGM
RUO
RNASEN
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications