Learn More
Abnova™ Human RNASEN Partial ORF (NP_037367.2, 1099 a.a. - 1196 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00029102-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Members of the ribonuclease III superfamily of double-stranded (ds) RNA-specific endoribonucleases participate in diverse RNA maturation and decay pathways in eukaryotic and prokaryotic cells (Fortin et al., 2002 [PubMed 12191433]). The RNase III Drosha is the core nuclease that executes the initiation step of microRNA (miRNA) processing in the nucleus (Lee et al., 2003 [PubMed 14508493]).[supplied by OMIM]
Sequence: ETSPVLQKLTEFEEAIGVIFTHVRLLARAFTLRTVGFNHLTLGHNQRMEFLGDSIMQLVATEYLFIHFPDHHEGHLTLLRSSLVNNRTQAKVAEELGMSpecifications
NP_037367.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ETSPVLQKLTEFEEAIGVIFTHVRLLARAFTLRTVGFNHLTLGHNQRMEFLGDSIMQLVATEYLFIHFPDHHEGHLTLLRSSLVNNRTQAKVAEELGM | |
RUO | |
RNASEN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
29102 | |
RNASEN (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DROSHA/ETOHI2/HSA242976/RANSE3L/RN3/RNASE3L | |
RNASEN | |
Recombinant | |
wheat germ expression system |