Description
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.
Specifications
Specifications
| Description | Human Reverse HPLC Primer, 50 nmole, For PCR and Sanger Sequencing |
| Product Type | Human Reverse HPLC Primer |
| Primer | Reverse |
| Quantity | 50 nmol |
| Grade | HPLC |