Learn More
STEMCELL Technologies Human Recombinant R-Spondin-1
Click to view available options
Quantity:
100 μg
1000 μg
25 μg
500 μg
Unit Size:
1000µg
100µg
25µg
500µg
Description
- R-Spondin-1 (RSPO1) is the prototype member of the R-Spondin (RSPO) protein subfamily of a superfamily of thrombospondin type 1 repeat (TSR-1)-containing proteins
- Although unable to initialize signaling, RSPO family members are potent enhancers of WNT signaling
- They are characterized by a TSR-1 domain, a carboxy-terminal region with positively charged amino acids, and two N-terminal furin-like cysteine-rich repeats
- R-Spondin-1 activates β-catenin signaling via the WNT signaling cascade and by indirectly increasing low-density lipoprotein receptor-related protein (LRP6) on the cell surface
Specifications
Specifications
| Name | hRspo1, Roof plate-specific Spondin-1 |
| Quantity | 1000 μg |
| Cell Type | Endoderm, PSC-Derived; Epithelial Cells; Hematopoietic Stem and Progenitor Cells; Intestinal Cells; Mesoderm, PSC-Derived; Other; Pluripotent Stem Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.