Learn More
STEMCELL Technologies Human Recombinant R-Spondin-1
Product Code. 17109861
Encompass_Preferred
Click to view available options
Quantity:
100 μg
1000 μg
25 μg
500 μg
Unit Size:
1000µg
100µg
25µg
500µg
Product Code. 17109861
Brand: STEMCELL Technologies 78213.3
R-Spondin-1
  • R-Spondin-1 (RSPO1) is the prototype member of the R-Spondin (RSPO) protein subfamily of a superfamily of thrombospondin type 1 repeat (TSR-1)-containing proteins
  • Although unable to initialize signaling, RSPO family members are potent enhancers of WNT signaling
  • They are characterized by a TSR-1 domain, a carboxy-terminal region with positively charged amino acids, and two N-terminal furin-like cysteine-rich repeats
  • R­-Spondin-1 activates β­-catenin signaling via the WNT signaling cascade and by indirectly increasing low-density lipoprotein receptor-related protein (LRP6) on the cell surface
Specifications
Name hRspo1, Roof plate-specific Spondin-1
Quantity 1000 μg
Cell Type Endoderm, PSC-Derived; Epithelial Cells; Hematopoietic Stem and Progenitor Cells; Intestinal Cells; Mesoderm, PSC-Derived; Other; Pluripotent Stem Cells
Purity or Quality Grade ≥95%