Learn More
STEMCELL Technologies Human Recombinant bFGF
Click to view available options
Quantity:
10 μg
1000 μg
50 μg
Unit Size:
1000µg
10µg
50µg
Description
- Basic fibroblast growth factor (bFGF) is a prototypic member of the fibroblast growth factor family
- Cytokines in the FGF family possess broad mitogenic and cell survival activities (Folkman & Klagsbrun; Kimelman & Kirschner) and are involved in a variety of biological processes including cell proliferation, differentiation, survival, and apoptosis (Folkman & Klagsbrun; Klagsbrun; Rifkin & Moscatelli)
- bFGF has the β-trefoil structure (Ponting & Russell), binds to the four FGF receptor (FGFR) family members, and activates JAK/STAT, PI3K, ERK1/2, and other receptor tyrosine kinase (RTK) signaling pathways
- It supports the maintenance of undifferentiated human embryonic stem cells (Xu et al.; Kang et al.), stimulates human embryonic stem cells to form neural rosettes (Zhang et al.), and improves proliferation of human mesenchymal stem cells and enhances chondrogenic differentiation (Solchaga et al.)
Specifications
Specifications
| Name | Basic fibroblast growth factor, FGF-β, FGF2, Fibroblast growth factor-basic, HBGF-2 |
| Quantity | 50 μg |
| Cell Type | Brain Tumor Stem Cells; Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Pluripotent Stem Cells; Prostate Cells |
| Purity or Quality Grade | >95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.