missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RARRES1 Full-length ORF (NP_002879.2, 1 a.a. - 228 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005918-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16164125

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene was identified as a retinoid acid (RA) receptor-responsive gene. It encodes a type 1 membrane protein. The expression of this gene is upregulated by tazarotene as well as by retinoic acid receptors. The expression of this gene is found to be downregulated in prostate cancer, which is caused by the methylation of its promoter and CpG island. Alternatively spliced transcript variant encoding distinct isoforms have been observed. [provided by RefSeq]

Sequence: MQPRRQRLPAPWSGPRGPRPTAPLLALLLLLAPVAAPAGSGDPDDPGQPQDAGVPRRLLQQAARAALHFFNFRSGSPSALRVLAEVQEGRAWINPKEGCKVHVVFSTERYNPESLLQEGEGRLGKCSARVFFKNQKPRPTINVTCTRLIEKKKRQQEDYLLYKQMKQLKNPLEIVSIPDNHGHIDPSLRLIWDLAFLGSSYVMWEMTTQVSHYYLAQLTSVRQWVRKT
Specifications

Specifications

NP_002879.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50.71kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TIG1
RARRES1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5918
RARRES1 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue
MQPRRQRLPAPWSGPRGPRPTAPLLALLLLLAPVAAPAGSGDPDDPGQPQDAGVPRRLLQQAARAALHFFNFRSGSPSALRVLAEVQEGRAWINPKEGCKVHVVFSTERYNPESLLQEGEGRLGKCSARVFFKNQKPRPTINVTCTRLIEKKKRQQEDYLLYKQMKQLKNPLEIVSIPDNHGHIDPSLRLIWDLAFLGSSYVMWEMTTQVSHYYLAQLTSVRQWVRKT
RUO
RARRES1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers