missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RARRES1 Full-length ORF (NP_002879.2, 1 a.a. - 228 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_002879.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5918 |
Molecular Weight (g/mol) | 50.71kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16174125
|
Abnova™
H00005918-P02.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16164125
|
Abnova™
H00005918-P02.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene was identified as a retinoid acid (RA) receptor-responsive gene. It encodes a type 1 membrane protein. The expression of this gene is upregulated by tazarotene as well as by retinoic acid receptors. The expression of this gene is found to be downregulated in prostate cancer, which is caused by the methylation of its promoter and CpG island. Alternatively spliced transcript variant encoding distinct isoforms have been observed. [provided by RefSeq]
Sequence: MQPRRQRLPAPWSGPRGPRPTAPLLALLLLLAPVAAPAGSGDPDDPGQPQDAGVPRRLLQQAARAALHFFNFRSGSPSALRVLAEVQEGRAWINPKEGCKVHVVFSTERYNPESLLQEGEGRLGKCSARVFFKNQKPRPTINVTCTRLIEKKKRQQEDYLLYKQMKQLKNPLEIVSIPDNHGHIDPSLRLIWDLAFLGSSYVMWEMTTQVSHYYLAQLTSVRQWVRKTSpecifications
NP_002879.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.71kDa | |
Glutathione Sepharose 4 Fast Flow | |
MQPRRQRLPAPWSGPRGPRPTAPLLALLLLLAPVAAPAGSGDPDDPGQPQDAGVPRRLLQQAARAALHFFNFRSGSPSALRVLAEVQEGRAWINPKEGCKVHVVFSTERYNPESLLQEGEGRLGKCSARVFFKNQKPRPTINVTCTRLIEKKKRQQEDYLLYKQMKQLKNPLEIVSIPDNHGHIDPSLRLIWDLAFLGSSYVMWEMTTQVSHYYLAQLTSVRQWVRKT | |
RUO | |
RARRES1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
5918 | |
RARRES1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TIG1 | |
RARRES1 | |
Recombinant | |
wheat germ expression system |