missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RAB25 Full-length ORF (NP_065120.2, 1 a.a. - 213 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057111-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16152493

  • 335.00€ / 10µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

RAB proteins, such as RAB25, are members of the RAS superfamily of small GTPases that are involved in membrane trafficking. Members of the RAB11 subfamily, including RAB25, control the return of internalized membrane-associated moieties to the cell surface (Caswell et al., 2007 [PubMed 17925226]).[supplied by OMIM]

Sequence: MGNGTEEDYNFVFKVVLIGESGVGKTNLLSRFTRNEFSHDSRTTIGVEFSTRTVMLGTAAVKAQIWDTAGLERYRAITSAYYRGAVGALLVFDLTKHQTYAVVERWLKELYDHAEATIVVMLVGNKSDLSQAREVPTEEARMFAENNGLLFLETSALDSTNVELAFETVLKEIFAKVSKQRQNSIRTNAITLGSAQAGQEPGPGEKRACCISL
Specifications

Specifications

NP_065120.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CATX-8
RAB25
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
57111
RAB25 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGNGTEEDYNFVFKVVLIGESGVGKTNLLSRFTRNEFSHDSRTTIGVEFSTRTVMLGTAAVKAQIWDTAGLERYRAITSAYYRGAVGALLVFDLTKHQTYAVVERWLKELYDHAEATIVVMLVGNKSDLSQAREVPTEEARMFAENNGLLFLETSALDSTNVELAFETVLKEIFAKVSKQRQNSIRTNAITLGSAQAGQEPGPGEKRACCISL
RUO
RAB25
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers