missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RAB25 Full-length ORF (NP_065120.2, 1 a.a. - 213 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_065120.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 57111 |
Molecular Weight (g/mol) | 49.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16152493
|
Abnova™
H00057111-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
16162493
|
Abnova™
H00057111-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
RAB proteins, such as RAB25, are members of the RAS superfamily of small GTPases that are involved in membrane trafficking. Members of the RAB11 subfamily, including RAB25, control the return of internalized membrane-associated moieties to the cell surface (Caswell et al., 2007 [PubMed 17925226]).[supplied by OMIM]
Sequence: MGNGTEEDYNFVFKVVLIGESGVGKTNLLSRFTRNEFSHDSRTTIGVEFSTRTVMLGTAAVKAQIWDTAGLERYRAITSAYYRGAVGALLVFDLTKHQTYAVVERWLKELYDHAEATIVVMLVGNKSDLSQAREVPTEEARMFAENNGLLFLETSALDSTNVELAFETVLKEIFAKVSKQRQNSIRTNAITLGSAQAGQEPGPGEKRACCISLSpecifications
NP_065120.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGNGTEEDYNFVFKVVLIGESGVGKTNLLSRFTRNEFSHDSRTTIGVEFSTRTVMLGTAAVKAQIWDTAGLERYRAITSAYYRGAVGALLVFDLTKHQTYAVVERWLKELYDHAEATIVVMLVGNKSDLSQAREVPTEEARMFAENNGLLFLETSALDSTNVELAFETVLKEIFAKVSKQRQNSIRTNAITLGSAQAGQEPGPGEKRACCISL | |
RUO | |
RAB25 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
57111 | |
RAB25 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CATX-8 | |
RAB25 | |
Recombinant | |
wheat germ expression system |