Learn More
ACROBiosystems Human R-Spondin 1 / RSPO1 Protein, His Tag
Brand: ACROBiosystems RS1-H4221-1MG
Additional Details : Weight : 0.00001kg
Description
R-spondin-1 is also known as Roof plate-specific Spondin 1 (RSPO1) and cysteinerich and single thrombospondin domain containing protein 3 (Cristin 3), which is a secreted protein which belongs to the R-Spondin family and encodes a secreted activator protein with two cystein-rich, furin-like domains and one thrombospondin type 1 domain. All Rspondins regulate Wnt/β-catenin signaling, but have distinct expression patterns. Like other R-Spondins, R-Spondin-1 contains two adjacent cysteinerich furinlike domains (aa 34-135) with one potential N-glycosylation site, followed by a thrombospondin (TSP1) motif (aa 147-207) and a region rich in basic residues (aa 211-263). Only the furinlike domains are needed for β-catenin stabilization. A putative nuclear localization signal at the C-terminus may allow some expression in the nucleus.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.7 kDa. The protein migrates as 38-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_001033722.1 | |
1 mg | |
Ser 21 - Ala 263 | |
RSPO1,CRISTIN3,FLJ40906,RP11-566C13.1,R-spondin-1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
28.7 kDa | |
Human R-Spondin 1, His Tag (RS1-H4221) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ala 263 (Accession # Q2MKA7-1). | |
-20°C | |
R-Spondin 1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |