missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human R-Spondin 1 / RSPO1 Protein, His Tag

457.25€ - 4350.40€

Specifications

Accession Number NP_001033722.1
Molecular Weight (g/mol) 28.7 kDa
Source Human R-Spondin 1, His Tag (RS1-H4221) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ala 263 (Accession # Q2MKA7-1).
Immunogen Ser 21 - Ala 263
Storage Requirements -20°C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15817627
View Documents Promotion Details
ACROBiosystems
RS1-H4221-50UG
50 μg
457.25€
50µg
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
 
15807627
View Documents Promotion Details
ACROBiosystems
RS1-H4221-1MG
1 mg
4350.40€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
 
Description

Description

R-spondin-1 is also known as Roof plate-specific Spondin 1 (RSPO1) and cysteinerich and single thrombospondin domain containing protein 3 (Cristin 3), which is a secreted protein which belongs to the R-Spondin family and encodes a secreted activator protein with two cystein-rich, furin-like domains and one thrombospondin type 1 domain. All Rspondins regulate Wnt/β-catenin signaling, but have distinct expression patterns. Like other R-Spondins, R-Spondin-1 contains two adjacent cysteinerich furinlike domains (aa 34-135) with one potential N-glycosylation site, followed by a thrombospondin (TSP1) motif (aa 147-207) and a region rich in basic residues (aa 211-263). Only the furinlike domains are needed for β-catenin stabilization. A putative nuclear localization signal at the C-terminus may allow some expression in the nucleus.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.7 kDa. The protein migrates as 38-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

NP_001033722.1
Human R-Spondin 1, His Tag (RS1-H4221) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ala 263 (Accession # Q2MKA7-1).
-20°C
R-Spondin 1
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens
28.7 kDa
Ser 21 - Ala 263
RSPO1,CRISTIN3,FLJ40906,RP11-566C13.1,R-spondin-1
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Videos
SDS
Documents

Documents

Product Certifications