missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PXR/NR1I2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems PP-H4417-00
This item is not returnable.
View return policy
Description
PXR/NR1I2 Monoclonal specifically detects PXR/NR1I2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Direct ELISA.
Specifications
| PXR/NR1I2 | |
| Monoclonal | |
| Unconjugated | |
| A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide | |
| NR1I2 | |
| Recombinant human PXR/NR1I2 | |
| 100 μg | |
| Primary | |
| This antibody specifically recognizes human PXR1 and human PXR2. Not yet tested in other species. | |
| This antibody is stable for greater than six months when stored at -20 degreesC in a manual defrost freezer or at -70 degreesC. Upon thawing, the antibody can be stored at 2-8 degreesC for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunoprecipitation, ELISA | |
| H4417 | |
| Western Blot, Immunoprecipitation, Direct ELISA | |
| BXR, NR1I2, nuclear receptor subfamily 1 group I member 2, nuclear receptor subfamily 1, group I, member 2, ONR1, Orphan nuclear receptor PAR1, Orphan nuclear receptor PXR, PAR, PAR1, PAR2, Pregnane X receptor, PRR, PXRPARq, SAR, Steroid and xenobiotic receptor, SXR, SXRBXR | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8856 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction