Learn More
Abnova™ Human POPDC3 Full-length ORF (NP_071756.2, 1 a.a. - 291 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064208-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the POP family of proteins containing three putative transmembrane domains. This gene is expressed in cardiac and skeletal muscle and may play an important role in these tissues during development. Alternatively spliced transcript variants have been found. [provided by RefSeq]
Sequence: MERNSSLWKNLIDEHPVCTTWKQEAEGAIYHLASILFVVGFMGGSGFFGLLYVFSLLGLGFLCSAVWAWVDVCAADIFSWNFVLFVICFMQFVHIAYQVRSITFAREFQVLYSSLFQPLGISLPVFRTIALSSEVVTLEKEHCYAMQGKTSIDKLSLLVSGRIRVTVDGEFLHYIFPLQFLDSPEWDSLRPTEEGIFQVTLTAETDCRYVSWRRKKLYLLFAQHRYISRLFSVLIGSDIADKLYALNDRVYIGKRYHYDIRLPNFYQMSTPEIRRSPLTQHFQNSRRYCDKSpecifications
NP_071756.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC22671/POP3/RP11-99L11.2/bA355M14.1 | |
POPDC3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
64208 | |
POPDC3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MERNSSLWKNLIDEHPVCTTWKQEAEGAIYHLASILFVVGFMGGSGFFGLLYVFSLLGLGFLCSAVWAWVDVCAADIFSWNFVLFVICFMQFVHIAYQVRSITFAREFQVLYSSLFQPLGISLPVFRTIALSSEVVTLEKEHCYAMQGKTSIDKLSLLVSGRIRVTVDGEFLHYIFPLQFLDSPEWDSLRPTEEGIFQVTLTAETDCRYVSWRRKKLYLLFAQHRYISRLFSVLIGSDIADKLYALNDRVYIGKRYHYDIRLPNFYQMSTPEIRRSPLTQHFQNSRRYCDK | |
RUO | |
POPDC3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |