missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human POPDC3 Full-length ORF (NP_071756.2, 1 a.a. - 291 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_071756.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 64208 |
Molecular Weight (g/mol) | 60.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16133473
|
Abnova™
H00064208-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16123473
|
Abnova™
H00064208-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the POP family of proteins containing three putative transmembrane domains. This gene is expressed in cardiac and skeletal muscle and may play an important role in these tissues during development. Alternatively spliced transcript variants have been found. [provided by RefSeq]
Sequence: MERNSSLWKNLIDEHPVCTTWKQEAEGAIYHLASILFVVGFMGGSGFFGLLYVFSLLGLGFLCSAVWAWVDVCAADIFSWNFVLFVICFMQFVHIAYQVRSITFAREFQVLYSSLFQPLGISLPVFRTIALSSEVVTLEKEHCYAMQGKTSIDKLSLLVSGRIRVTVDGEFLHYIFPLQFLDSPEWDSLRPTEEGIFQVTLTAETDCRYVSWRRKKLYLLFAQHRYISRLFSVLIGSDIADKLYALNDRVYIGKRYHYDIRLPNFYQMSTPEIRRSPLTQHFQNSRRYCDKSpecifications
NP_071756.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MERNSSLWKNLIDEHPVCTTWKQEAEGAIYHLASILFVVGFMGGSGFFGLLYVFSLLGLGFLCSAVWAWVDVCAADIFSWNFVLFVICFMQFVHIAYQVRSITFAREFQVLYSSLFQPLGISLPVFRTIALSSEVVTLEKEHCYAMQGKTSIDKLSLLVSGRIRVTVDGEFLHYIFPLQFLDSPEWDSLRPTEEGIFQVTLTAETDCRYVSWRRKKLYLLFAQHRYISRLFSVLIGSDIADKLYALNDRVYIGKRYHYDIRLPNFYQMSTPEIRRSPLTQHFQNSRRYCDK | |
RUO | |
POPDC3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
64208 | |
POPDC3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC22671/POP3/RP11-99L11.2/bA355M14.1 | |
POPDC3 | |
Recombinant | |
wheat germ expression system |