missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human POPDC3 Full-length ORF (NP_071756.2, 1 a.a. - 291 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00064208-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16123473

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the POP family of proteins containing three putative transmembrane domains. This gene is expressed in cardiac and skeletal muscle and may play an important role in these tissues during development. Alternatively spliced transcript variants have been found. [provided by RefSeq]

Sequence: MERNSSLWKNLIDEHPVCTTWKQEAEGAIYHLASILFVVGFMGGSGFFGLLYVFSLLGLGFLCSAVWAWVDVCAADIFSWNFVLFVICFMQFVHIAYQVRSITFAREFQVLYSSLFQPLGISLPVFRTIALSSEVVTLEKEHCYAMQGKTSIDKLSLLVSGRIRVTVDGEFLHYIFPLQFLDSPEWDSLRPTEEGIFQVTLTAETDCRYVSWRRKKLYLLFAQHRYISRLFSVLIGSDIADKLYALNDRVYIGKRYHYDIRLPNFYQMSTPEIRRSPLTQHFQNSRRYCDK
Specifications

Specifications

NP_071756.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
60.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC22671/POP3/RP11-99L11.2/bA355M14.1
POPDC3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
64208
POPDC3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MERNSSLWKNLIDEHPVCTTWKQEAEGAIYHLASILFVVGFMGGSGFFGLLYVFSLLGLGFLCSAVWAWVDVCAADIFSWNFVLFVICFMQFVHIAYQVRSITFAREFQVLYSSLFQPLGISLPVFRTIALSSEVVTLEKEHCYAMQGKTSIDKLSLLVSGRIRVTVDGEFLHYIFPLQFLDSPEWDSLRPTEEGIFQVTLTAETDCRYVSWRRKKLYLLFAQHRYISRLFSVLIGSDIADKLYALNDRVYIGKRYHYDIRLPNFYQMSTPEIRRSPLTQHFQNSRRYCDK
RUO
POPDC3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers