missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PMPCB Full-length ORF (AAH10398.1, 1 a.a. - 490 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009512-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193752

  • 349.00€ / 10µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

This gene is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex. [provided by RefSeq]

Sequence: MAAAAARVVLSSAARRRLWGFSESLLIRGAAGRSLYFGENRLRSTQAATQVVLNVPETRVTCLESGLRVASEDSGLSTCTVGLWIDAGSRYENEKNNGTAHFLEHMAFKGTKKRSQLDLELEIENMGAHLNAYTSREQTVYYAKAFSKDLPRAVEILADIIQNSTLGEAEIERERGVILREMQEVETNLQEVVFDYLHATAYQNTALGRTILGPTENIKSISRKDLVDYITTHYKGPRIVLAAAGGVSHDELLDLAKFHFGDSLCTHKGEIPALPPCKFTGSEIRVRDDKMPLAHLAIAVEAVGWAHPDTICLMVANTLIGNWDRSFGGGMNLSSKLAQLTCHGNLCHSFQSFNTSYTDTGLWGLYMVCESSTVADMLHVVQKEWMRLCTSVTESEVARARNLLKTNMLLQLDGSTPICEDIGRQMLCYNRRIPIPELEARIDAVNAETIREVCTKYIYNRSPAIAAVGKPGFFSSMQKVGQVLLINSSF
Specifications

Specifications

AAH10398.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
80.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Beta-MPP/MPP11/MPPB/MPPP52
PMPCB
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9512
PMPCB (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAAAARVVLSSAARRRLWGFSESLLIRGAAGRSLYFGENRLRSTQAATQVVLNVPETRVTCLESGLRVASEDSGLSTCTVGLWIDAGSRYENEKNNGTAHFLEHMAFKGTKKRSQLDLELEIENMGAHLNAYTSREQTVYYAKAFSKDLPRAVEILADIIQNSTLGEAEIERERGVILREMQEVETNLQEVVFDYLHATAYQNTALGRTILGPTENIKSISRKDLVDYITTHYKGPRIVLAAAGGVSHDELLDLAKFHFGDSLCTHKGEIPALPPCKFTGSEIRVRDDKMPLAHLAIAVEAVGWAHPDTICLMVANTLIGNWDRSFGGGMNLSSKLAQLTCHGNLCHSFQSFNTSYTDTGLWGLYMVCESSTVADMLHVVQKEWMRLCTSVTESEVARARNLLKTNMLLQLDGSTPICEDIGRQMLCYNRRIPIPELEARIDAVNAETIREVCTKYIYNRSPAIAAVGKPGFFSSMQKVGQVLLINSSF
RUO
PMPCB
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers