missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PMPCB Full-length ORF (AAH10398.1, 1 a.a. - 490 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH10398.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9512 |
Molecular Weight (g/mol) | 80.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16103762
|
Abnova™
H00009512-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16193752
|
Abnova™
H00009512-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
This gene is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex. [provided by RefSeq]
Sequence: MAAAAARVVLSSAARRRLWGFSESLLIRGAAGRSLYFGENRLRSTQAATQVVLNVPETRVTCLESGLRVASEDSGLSTCTVGLWIDAGSRYENEKNNGTAHFLEHMAFKGTKKRSQLDLELEIENMGAHLNAYTSREQTVYYAKAFSKDLPRAVEILADIIQNSTLGEAEIERERGVILREMQEVETNLQEVVFDYLHATAYQNTALGRTILGPTENIKSISRKDLVDYITTHYKGPRIVLAAAGGVSHDELLDLAKFHFGDSLCTHKGEIPALPPCKFTGSEIRVRDDKMPLAHLAIAVEAVGWAHPDTICLMVANTLIGNWDRSFGGGMNLSSKLAQLTCHGNLCHSFQSFNTSYTDTGLWGLYMVCESSTVADMLHVVQKEWMRLCTSVTESEVARARNLLKTNMLLQLDGSTPICEDIGRQMLCYNRRIPIPELEARIDAVNAETIREVCTKYIYNRSPAIAAVGKPGFFSSMQKVGQVLLINSSFSpecifications
AAH10398.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
80.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAAAAARVVLSSAARRRLWGFSESLLIRGAAGRSLYFGENRLRSTQAATQVVLNVPETRVTCLESGLRVASEDSGLSTCTVGLWIDAGSRYENEKNNGTAHFLEHMAFKGTKKRSQLDLELEIENMGAHLNAYTSREQTVYYAKAFSKDLPRAVEILADIIQNSTLGEAEIERERGVILREMQEVETNLQEVVFDYLHATAYQNTALGRTILGPTENIKSISRKDLVDYITTHYKGPRIVLAAAGGVSHDELLDLAKFHFGDSLCTHKGEIPALPPCKFTGSEIRVRDDKMPLAHLAIAVEAVGWAHPDTICLMVANTLIGNWDRSFGGGMNLSSKLAQLTCHGNLCHSFQSFNTSYTDTGLWGLYMVCESSTVADMLHVVQKEWMRLCTSVTESEVARARNLLKTNMLLQLDGSTPICEDIGRQMLCYNRRIPIPELEARIDAVNAETIREVCTKYIYNRSPAIAAVGKPGFFSSMQKVGQVLLINSSF | |
RUO | |
PMPCB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9512 | |
PMPCB (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Beta-MPP/MPP11/MPPB/MPPP52 | |
PMPCB | |
Recombinant | |
wheat germ expression system |