Learn More
Abnova™ Human PMPCB Full-length ORF (AAH10398.1, 1 a.a. - 490 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009512-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex. [provided by RefSeq]
Sequence: MAAAAARVVLSSAARRRLWGFSESLLIRGAAGRSLYFGENRLRSTQAATQVVLNVPETRVTCLESGLRVASEDSGLSTCTVGLWIDAGSRYENEKNNGTAHFLEHMAFKGTKKRSQLDLELEIENMGAHLNAYTSREQTVYYAKAFSKDLPRAVEILADIIQNSTLGEAEIERERGVILREMQEVETNLQEVVFDYLHATAYQNTALGRTILGPTENIKSISRKDLVDYITTHYKGPRIVLAAAGGVSHDELLDLAKFHFGDSLCTHKGEIPALPPCKFTGSEIRVRDDKMPLAHLAIAVEAVGWAHPDTICLMVANTLIGNWDRSFGGGMNLSSKLAQLTCHGNLCHSFQSFNTSYTDTGLWGLYMVCESSTVADMLHVVQKEWMRLCTSVTESEVARARNLLKTNMLLQLDGSTPICEDIGRQMLCYNRRIPIPELEARIDAVNAETIREVCTKYIYNRSPAIAAVGKPGFFSSMQKVGQVLLINSSFSpecifications
AAH10398.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
80.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Beta-MPP/MPP11/MPPB/MPPP52 | |
PMPCB | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9512 | |
PMPCB (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAAAARVVLSSAARRRLWGFSESLLIRGAAGRSLYFGENRLRSTQAATQVVLNVPETRVTCLESGLRVASEDSGLSTCTVGLWIDAGSRYENEKNNGTAHFLEHMAFKGTKKRSQLDLELEIENMGAHLNAYTSREQTVYYAKAFSKDLPRAVEILADIIQNSTLGEAEIERERGVILREMQEVETNLQEVVFDYLHATAYQNTALGRTILGPTENIKSISRKDLVDYITTHYKGPRIVLAAAGGVSHDELLDLAKFHFGDSLCTHKGEIPALPPCKFTGSEIRVRDDKMPLAHLAIAVEAVGWAHPDTICLMVANTLIGNWDRSFGGGMNLSSKLAQLTCHGNLCHSFQSFNTSYTDTGLWGLYMVCESSTVADMLHVVQKEWMRLCTSVTESEVARARNLLKTNMLLQLDGSTPICEDIGRQMLCYNRRIPIPELEARIDAVNAETIREVCTKYIYNRSPAIAAVGKPGFFSSMQKVGQVLLINSSF | |
RUO | |
PMPCB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |