missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-Src (Y416) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
240.00€ - 517.00€
Specifications
| Antigen | Src |
|---|---|
| Clone | 1246F |
| Dilution | Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 1-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18787499
![]() |
R&D Systems
MAB2685-SP |
25 μg |
240.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18772157
![]() |
R&D Systems
MAB2685 |
100 μg |
517.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Src Monoclonal specifically detects Src in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
| Src | |
| Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 1-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| 6714 | |
| Phosphopeptide containing the human Src Y416 site | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Src when phosphorylated at Y416 in direct ELISAs and Western blots. |
| 1246F | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ASV, c-Src, EC 2.7.10, EC 2.7.10.2, pp60c-src, Rous sarcoma, RSVgp4, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian) | |
| SRC | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title