missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PHF2 Partial ORF (NP_005383, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005253-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16172735

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a protein which contains a zinc finger-like PHD (plant homeodomain) finger, distinct from other classes of zinc finger motifs, and a hydrophobic and highly conserved domain. The PHD finger shows the typical Cys4-His-Cys3 arrangement. PHD finger genes are thought to belong to a diverse group of transcriptional regulators possibly affecting eukaryotic gene expression by influencing chromatin structure. [provided by RefSeq]

Sequence: ATVPVYCVCRLPYDVTRFMIECDACKDWFHGSCVGVEEEEAPDIDIYHCPNCEKTHGKSTLKKKRTWHKHGPGQAPDVKPVQNGSQLFIKELRSRTFPS
Specifications

Specifications

NP_005383
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ATVPVYCVCRLPYDVTRFMIECDACKDWFHGSCVGVEEEEAPDIDIYHCPNCEKTHGKSTLKKKRTWHKHGPGQAPDVKPVQNGSQLFIKELRSRTFPS
RUO
PHF2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5253
PHF2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GRC5/JHDM1E/KIAA0662/MGC176680
PHF2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers