missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PHF2 Partial ORF (NP_005383, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_005383 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5253 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16162735
|
Abnova™
H00005253-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16172735
|
Abnova™
H00005253-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a protein which contains a zinc finger-like PHD (plant homeodomain) finger, distinct from other classes of zinc finger motifs, and a hydrophobic and highly conserved domain. The PHD finger shows the typical Cys4-His-Cys3 arrangement. PHD finger genes are thought to belong to a diverse group of transcriptional regulators possibly affecting eukaryotic gene expression by influencing chromatin structure. [provided by RefSeq]
Sequence: ATVPVYCVCRLPYDVTRFMIECDACKDWFHGSCVGVEEEEAPDIDIYHCPNCEKTHGKSTLKKKRTWHKHGPGQAPDVKPVQNGSQLFIKELRSRTFPSSpecifications
NP_005383 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GRC5/JHDM1E/KIAA0662/MGC176680 | |
PHF2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5253 | |
PHF2 (Human) Recombinant Protein (Q01) | |
ATVPVYCVCRLPYDVTRFMIECDACKDWFHGSCVGVEEEEAPDIDIYHCPNCEKTHGKSTLKKKRTWHKHGPGQAPDVKPVQNGSQLFIKELRSRTFPS | |
RUO | |
PHF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |