missing translation for 'onlineSavingsMsg'
Learn More

Human PGLYRP4/PGRP-I beta Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18031112
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18031112 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18031112 Supplier R&D Systems Supplier No. BAF3018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

PGLYRP4/PGRP-I beta Polyclonal specifically detects PGLYRP4/PGRP-I beta in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGLYRP4/PGRP-I beta
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. AAI07158
Gene Alias peptidoglycan recognition protein 4, peptidoglycan recognition protein I beta, Pglyrp-1 beta, PGRPI beta, PGRP-I beta, PGRP-I-beta
Gene Symbols PGLYRP4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human PGLYRP4/PGRP-I beta Asp18-Lys372 Accession # AAI07158
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57115
Test Specificity Detects human PGLYRP4/PGRP-I beta in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse PGRP-I beta is observed and 10% cross-reactivity with recombinant human PGRP-S is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.