Learn More
Abnova™ Human PELI3 Full-length ORF (AAH21256.1, 1 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00246330-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Toll-like receptors (TLRs; see MIM 603030) and IL1R (IL1R1; MIM 147810) are part of the innate immune response aimed at mobilizing defense mechanisms in response in infection or injury. Pellino proteins, such as PELI3, are intermediate components in the signaling cascades initiated by TLRs and IL1R (Jensen and Whitehead, 2003 [PubMed 12874243]).[supplied by OMIM]
Sequence: MVLEGNPEVGSPRTSDLQHRGNKGSCVLSSPGEDAQPGEEPIKYGELIVLGCCEEGGEETEAQRGEVTGPRAHSCYNGCLASGDKGRRRSRLALSRRSHANGVKPDVMHHISTPLVSKALSNRGQHSISYTLSRSHSVIVEYTHDSDTDMFQIGRSTENMIDFVVTDTSPGGGAAEGPSAQSTISRYACRILCDRRPPYTARIYAAGFDASSNIFLGERAAKWRTPDGLMDGLTTNGVLVMHPAGGFSEDSAPGVWREISVCGNVYTLRDSRSAQQRGKLVGGPLHSPPLSLPGPHKLPIPLPKPGDDLQSFCPREPQQAPYQGIPGPGGSSpecifications
AAH21256.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
61.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC35521 | |
PELI3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
246330 | |
PELI3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVLEGNPEVGSPRTSDLQHRGNKGSCVLSSPGEDAQPGEEPIKYGELIVLGCCEEGGEETEAQRGEVTGPRAHSCYNGCLASGDKGRRRSRLALSRRSHANGVKPDVMHHISTPLVSKALSNRGQHSISYTLSRSHSVIVEYTHDSDTDMFQIGRSTENMIDFVVTDTSPGGGAAEGPSAQSTISRYACRILCDRRPPYTARIYAAGFDASSNIFLGERAAKWRTPDGLMDGLTTNGVLVMHPAGGFSEDSAPGVWREISVCGNVYTLRDSRSAQQRGKLVGGPLHSPPLSLPGPHKLPIPLPKPGDDLQSFCPREPQQAPYQGIPGPGGS | |
RUO | |
PELI3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |