missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PELI3 Full-length ORF (AAH21256.1, 1 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH21256.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 246330 |
Molecular Weight (g/mol) | 61.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16159563
|
Abnova™
H00246330-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16149563
|
Abnova™
H00246330-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Toll-like receptors (TLRs; see MIM 603030) and IL1R (IL1R1; MIM 147810) are part of the innate immune response aimed at mobilizing defense mechanisms in response in infection or injury. Pellino proteins, such as PELI3, are intermediate components in the signaling cascades initiated by TLRs and IL1R (Jensen and Whitehead, 2003 [PubMed 12874243]).[supplied by OMIM]
Sequence: MVLEGNPEVGSPRTSDLQHRGNKGSCVLSSPGEDAQPGEEPIKYGELIVLGCCEEGGEETEAQRGEVTGPRAHSCYNGCLASGDKGRRRSRLALSRRSHANGVKPDVMHHISTPLVSKALSNRGQHSISYTLSRSHSVIVEYTHDSDTDMFQIGRSTENMIDFVVTDTSPGGGAAEGPSAQSTISRYACRILCDRRPPYTARIYAAGFDASSNIFLGERAAKWRTPDGLMDGLTTNGVLVMHPAGGFSEDSAPGVWREISVCGNVYTLRDSRSAQQRGKLVGGPLHSPPLSLPGPHKLPIPLPKPGDDLQSFCPREPQQAPYQGIPGPGGSSpecifications
AAH21256.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
61.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MVLEGNPEVGSPRTSDLQHRGNKGSCVLSSPGEDAQPGEEPIKYGELIVLGCCEEGGEETEAQRGEVTGPRAHSCYNGCLASGDKGRRRSRLALSRRSHANGVKPDVMHHISTPLVSKALSNRGQHSISYTLSRSHSVIVEYTHDSDTDMFQIGRSTENMIDFVVTDTSPGGGAAEGPSAQSTISRYACRILCDRRPPYTARIYAAGFDASSNIFLGERAAKWRTPDGLMDGLTTNGVLVMHPAGGFSEDSAPGVWREISVCGNVYTLRDSRSAQQRGKLVGGPLHSPPLSLPGPHKLPIPLPKPGDDLQSFCPREPQQAPYQGIPGPGGS | |
RUO | |
PELI3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
246330 | |
PELI3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC35521 | |
PELI3 | |
Recombinant | |
wheat germ expression system |