missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PDCL2 Full-length ORF (NP_689614.2, 1 a.a. - 241 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00132954-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16117993

  • 335.00€ / 10µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the phosducin-like protein family and is a putative modulator of heterotrimeric G proteins. The protein shares extensive amino acid sequence homology with phosducin. Members of the phosducin-like protein family have been shown to bind to the beta-gamma subunits of G proteins. [provided by RefSeq]

Sequence: MQDPNEDTEWNDILRDFGILPPKEESKDEIEEMVLRLQKEAMVKPFEKMTLAQLKEAEDEFDEEDMQAVETYRKKRLQEWKALKKKQKFGELREISGNQYVNEVTNAEEDVWVIIHLYRSSIPMCLLVNQHLSLLARKFPETKFVKAIVNSCIQHYHDNCLPTIFVYKNGQIEAKFIGIIECGGINLKLEELEWKLAEVGAIQTDLEENPRKDMVDMMVSSIRNTSIHDDSDSSNSDNDTK
Specifications

Specifications

NP_689614.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
54.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GCPHLP
PDCL2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
132954
PDCL2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQDPNEDTEWNDILRDFGILPPKEESKDEIEEMVLRLQKEAMVKPFEKMTLAQLKEAEDEFDEEDMQAVETYRKKRLQEWKALKKKQKFGELREISGNQYVNEVTNAEEDVWVIIHLYRSSIPMCLLVNQHLSLLARKFPETKFVKAIVNSCIQHYHDNCLPTIFVYKNGQIEAKFIGIIECGGINLKLEELEWKLAEVGAIQTDLEENPRKDMVDMMVSSIRNTSIHDDSDSSNSDNDTK
RUO
PDCL2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers