missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PDCL2 Full-length ORF (NP_689614.2, 1 a.a. - 241 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_689614.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 132954 |
Molecular Weight (g/mol) | 54.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16127993
|
Abnova™
H00132954-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16117993
|
Abnova™
H00132954-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the phosducin-like protein family and is a putative modulator of heterotrimeric G proteins. The protein shares extensive amino acid sequence homology with phosducin. Members of the phosducin-like protein family have been shown to bind to the beta-gamma subunits of G proteins. [provided by RefSeq]
Sequence: MQDPNEDTEWNDILRDFGILPPKEESKDEIEEMVLRLQKEAMVKPFEKMTLAQLKEAEDEFDEEDMQAVETYRKKRLQEWKALKKKQKFGELREISGNQYVNEVTNAEEDVWVIIHLYRSSIPMCLLVNQHLSLLARKFPETKFVKAIVNSCIQHYHDNCLPTIFVYKNGQIEAKFIGIIECGGINLKLEELEWKLAEVGAIQTDLEENPRKDMVDMMVSSIRNTSIHDDSDSSNSDNDTKSpecifications
NP_689614.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MQDPNEDTEWNDILRDFGILPPKEESKDEIEEMVLRLQKEAMVKPFEKMTLAQLKEAEDEFDEEDMQAVETYRKKRLQEWKALKKKQKFGELREISGNQYVNEVTNAEEDVWVIIHLYRSSIPMCLLVNQHLSLLARKFPETKFVKAIVNSCIQHYHDNCLPTIFVYKNGQIEAKFIGIIECGGINLKLEELEWKLAEVGAIQTDLEENPRKDMVDMMVSSIRNTSIHDDSDSSNSDNDTK | |
RUO | |
PDCL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
132954 | |
PDCL2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GCPHLP | |
PDCL2 | |
Recombinant | |
wheat germ expression system |