missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PCDHGB2 Full-length ORF (NP_115267.1, 1 a.a. - 811 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00056103-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122053

  • 508.00€ / 25µg
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. [provided by RefSeq]

Sequence: MKASSGRCGLVRWLQVLLPFLLSLFPGALPVQIRYSIPEELAKNSVVGNLAKDLGLSVRDLPARKLRVSAEKEYFTVNPESGDLLVSDRIDREQICGKQPLCVLDFDTVAENPLNIFYIAVIVQDINDNTPLFKQTKINLKIGESTKPGTTFPLDPALDSDVGPNSLQRYHLNDNEYFDLAEKQTPDGRKYPELILKHSLDREEHSLHQLVLTAVDGGDPPQSGTTQIRIKVTDANDNPPVFSQDVYRVTLREDVPPGFFVLQVTATDRDEGINAEITYSFHNVDEQVKHFFNLNEKTGEITTKDDLDFEIASSYTLSIEAKDPGDLAAHCSIQVEILDDNDCAPEVIVTSVSTPLPEDSPPGTVIALIKTRDRDSGENGEVYCQVLGNAKFILKSSSKNYYKLVTDGALDREEIPEYNLTITATDGGKPPLSSSIIVTLHISDVNDNAPVFQQTSYMVHVAENNPPGASIAQISASDPDLGPSGQVSYSIVASDLKPREILSYVSVSAQSGVVFAQRAFDHEQLRAFELTLQARDQGSPALSANVSLRVLVGDLNDNAPRVLYPALGPDGSALFDMVPRAAEPGYLVTKVVAVDADSGHNAWLSYHVLQASEPGLFSLGLRTGEVRTARALGDRDAARQRLLVAVRDGGQPPLSATATLHLIFADSLQEVLPDLSDRREPSDPQAKLQFYLVVALALISVLFFLAVILAISLRLRLSSRSDAWDCFQPGLSSKPGPGVLPNYSEGTLPYSYNLCVASQSAKTEFNFLNITPELVPAQDLVCDNASWEQNTNHGAAGVPFASDTILKVSFN
Specifications

Specifications

NP_115267.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
114.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC126854/PCDH-GAMMA-B2
PCDHGB2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
56103
PCDHGB2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKASSGRCGLVRWLQVLLPFLLSLFPGALPVQIRYSIPEELAKNSVVGNLAKDLGLSVRDLPARKLRVSAEKEYFTVNPESGDLLVSDRIDREQICGKQPLCVLDFDTVAENPLNIFYIAVIVQDINDNTPLFKQTKINLKIGESTKPGTTFPLDPALDSDVGPNSLQRYHLNDNEYFDLAEKQTPDGRKYPELILKHSLDREEHSLHQLVLTAVDGGDPPQSGTTQIRIKVTDANDNPPVFSQDVYRVTLREDVPPGFFVLQVTATDRDEGINAEITYSFHNVDEQVKHFFNLNEKTGEITTKDDLDFEIASSYTLSIEAKDPGDLAAHCSIQVEILDDNDCAPEVIVTSVSTPLPEDSPPGTVIALIKTRDRDSGENGEVYCQVLGNAKFILKSSSKNYYKLVTDGALDREEIPEYNLTITATDGGKPPLSSSIIVTLHISDVNDNAPVFQQTSYMVHVAENNPPGASIAQISASDPDLGPSGQVSYSIVASDLKPREILSYVSVSAQSGVVFAQRAFDHEQLRAFELTLQARDQGSPALSANVSLRVLVGDLNDNAPRVLYPALGPDGSALFDMVPRAAEPGYLVTKVVAVDADSGHNAWLSYHVLQASEPGLFSLGLRTGEVRTARALGDRDAARQRLLVAVRDGGQPPLSATATLHLIFADSLQEVLPDLSDRREPSDPQAKLQFYLVVALALISVLFFLAVILAISLRLRLSSRSDAWDCFQPGLSSKPGPGVLPNYSEGTLPYSYNLCVASQSAKTEFNFLNITPELVPAQDLVCDNASWEQNTNHGAAGVPFASDTILKVSFN
RUO
PCDHGB2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers