Learn More
Abnova™ Human PCDHGB2 Full-length ORF (NP_115267.1, 1 a.a. - 811 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_115267.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 56103 |
Molecular Weight (g/mol) | 114.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16122053
|
Abnova™
H00056103-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16112053
|
Abnova™
H00056103-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. [provided by RefSeq]
Sequence: MKASSGRCGLVRWLQVLLPFLLSLFPGALPVQIRYSIPEELAKNSVVGNLAKDLGLSVRDLPARKLRVSAEKEYFTVNPESGDLLVSDRIDREQICGKQPLCVLDFDTVAENPLNIFYIAVIVQDINDNTPLFKQTKINLKIGESTKPGTTFPLDPALDSDVGPNSLQRYHLNDNEYFDLAEKQTPDGRKYPELILKHSLDREEHSLHQLVLTAVDGGDPPQSGTTQIRIKVTDANDNPPVFSQDVYRVTLREDVPPGFFVLQVTATDRDEGINAEITYSFHNVDEQVKHFFNLNEKTGEITTKDDLDFEIASSYTLSIEAKDPGDLAAHCSIQVEILDDNDCAPEVIVTSVSTPLPEDSPPGTVIALIKTRDRDSGENGEVYCQVLGNAKFILKSSSKNYYKLVTDGALDREEIPEYNLTITATDGGKPPLSSSIIVTLHISDVNDNAPVFQQTSYMVHVAENNPPGASIAQISASDPDLGPSGQVSYSIVASDLKPREILSYVSVSAQSGVVFAQRAFDHEQLRAFELTLQARDQGSPALSANVSLRVLVGDLNDNAPRVLYPALGPDGSALFDMVPRAAEPGYLVTKVVAVDADSGHNAWLSYHVLQASEPGLFSLGLRTGEVRTARALGDRDAARQRLLVAVRDGGQPPLSATATLHLIFADSLQEVLPDLSDRREPSDPQAKLQFYLVVALALISVLFFLAVILAISLRLRLSSRSDAWDCFQPGLSSKPGPGVLPNYSEGTLPYSYNLCVASQSAKTEFNFLNITPELVPAQDLVCDNASWEQNTNHGAAGVPFASDTILKVSFNSpecifications
NP_115267.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
114.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKASSGRCGLVRWLQVLLPFLLSLFPGALPVQIRYSIPEELAKNSVVGNLAKDLGLSVRDLPARKLRVSAEKEYFTVNPESGDLLVSDRIDREQICGKQPLCVLDFDTVAENPLNIFYIAVIVQDINDNTPLFKQTKINLKIGESTKPGTTFPLDPALDSDVGPNSLQRYHLNDNEYFDLAEKQTPDGRKYPELILKHSLDREEHSLHQLVLTAVDGGDPPQSGTTQIRIKVTDANDNPPVFSQDVYRVTLREDVPPGFFVLQVTATDRDEGINAEITYSFHNVDEQVKHFFNLNEKTGEITTKDDLDFEIASSYTLSIEAKDPGDLAAHCSIQVEILDDNDCAPEVIVTSVSTPLPEDSPPGTVIALIKTRDRDSGENGEVYCQVLGNAKFILKSSSKNYYKLVTDGALDREEIPEYNLTITATDGGKPPLSSSIIVTLHISDVNDNAPVFQQTSYMVHVAENNPPGASIAQISASDPDLGPSGQVSYSIVASDLKPREILSYVSVSAQSGVVFAQRAFDHEQLRAFELTLQARDQGSPALSANVSLRVLVGDLNDNAPRVLYPALGPDGSALFDMVPRAAEPGYLVTKVVAVDADSGHNAWLSYHVLQASEPGLFSLGLRTGEVRTARALGDRDAARQRLLVAVRDGGQPPLSATATLHLIFADSLQEVLPDLSDRREPSDPQAKLQFYLVVALALISVLFFLAVILAISLRLRLSSRSDAWDCFQPGLSSKPGPGVLPNYSEGTLPYSYNLCVASQSAKTEFNFLNITPELVPAQDLVCDNASWEQNTNHGAAGVPFASDTILKVSFN | |
RUO | |
PCDHGB2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
56103 | |
PCDHGB2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126854/PCDH-GAMMA-B2 | |
PCDHGB2 | |
Recombinant | |
wheat germ expression system |