missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Patched 2/PTCH2 (First Extracellular Loop) Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4078-SP
This item is not returnable.
View return policy
Description
Patched 2/PTCH2 Polyclonal specifically detects Patched 2/PTCH2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Patched 2/PTCH2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| patched (Drosophila) homolog 2, patched 2, patched homolog 2, patched homolog 2 (Drosophila), protein patched homolog 2, ptc2, PTCH2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8643 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q9Y6C5 | |
| PTCH2 | |
| E. coli-derived recombinant human Patched 2/PTCH2 first extracellular loop Glu79-Glu392 Accession # Q9Y6C5 | |
| 25 μg | |
| Primary | |
| Detects human Patched 2/PTCH2 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse Patched is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction