Learn More
Abnova™ Human PARD6A Full-length ORF (NP_001032358.1, 1 a.a. - 345 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00050855-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the PAR6 family and encodes a protein with a PSD95/Discs-large/ZO1 (PDZ) domain and a semi-Cdc42/Rac interactive binding (CRIB) domain. This cell membrane protein is involved in asymmetrical cell division and cell polarization processes as a member of a multi-protein complex. The protein also has a role in the epithelial-to-mesenchymal transition (EMT) that characterizes the invasive phenotype associated with metastatic carcinomas. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: MARPQRTPARSPDSIVEVKSKFDAEFRRFALPRASVSGFQEFSRLLRAVHQIPGLDVLLGYTDAHGDLLPLTNDDSLHRALASGPPPLRLLVQKREADSSGLAFASNSLQRRKKGLLLRPVAPLRTRPPLLISLPQDFRQVSSVIDVDLLPETHRRVRLHKHGSDRPLGFYIRDGMSVRVAPQGLERVPGIFISRLVRGGLAESTGLLAVSDEILEVNGIEVAGKTLDQVTDMMVANSHNLIVTVKPANQRNNVVRGASGRLTGPPSAGPGPAEPDSDDDSSDLVIENRQPPSSNGLSQGPPCWDLHPGCRHPGTRSSLPSLDDQEQASSGWGSRIRGDGSGFSLSpecifications
NP_001032358.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PAR-6A/PAR6/PAR6C/PAR6alpha/TAX40/TIP-40 | |
PARD6A | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
50855 | |
PARD6A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MARPQRTPARSPDSIVEVKSKFDAEFRRFALPRASVSGFQEFSRLLRAVHQIPGLDVLLGYTDAHGDLLPLTNDDSLHRALASGPPPLRLLVQKREADSSGLAFASNSLQRRKKGLLLRPVAPLRTRPPLLISLPQDFRQVSSVIDVDLLPETHRRVRLHKHGSDRPLGFYIRDGMSVRVAPQGLERVPGIFISRLVRGGLAESTGLLAVSDEILEVNGIEVAGKTLDQVTDMMVANSHNLIVTVKPANQRNNVVRGASGRLTGPPSAGPGPAEPDSDDDSSDLVIENRQPPSSNGLSQGPPCWDLHPGCRHPGTRSSLPSLDDQEQASSGWGSRIRGDGSGFSL | |
RUO | |
PARD6A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |