Learn More
Abnova™ Human PARD6A Full-length ORF (NP_001032358.1, 1 a.a. - 345 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_001032358.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 50855 |
Molecular Weight (g/mol) | 63.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16159452
|
Abnova™
H00050855-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16149452
|
Abnova™
H00050855-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene is a member of the PAR6 family and encodes a protein with a PSD95/Discs-large/ZO1 (PDZ) domain and a semi-Cdc42/Rac interactive binding (CRIB) domain. This cell membrane protein is involved in asymmetrical cell division and cell polarization processes as a member of a multi-protein complex. The protein also has a role in the epithelial-to-mesenchymal transition (EMT) that characterizes the invasive phenotype associated with metastatic carcinomas. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: MARPQRTPARSPDSIVEVKSKFDAEFRRFALPRASVSGFQEFSRLLRAVHQIPGLDVLLGYTDAHGDLLPLTNDDSLHRALASGPPPLRLLVQKREADSSGLAFASNSLQRRKKGLLLRPVAPLRTRPPLLISLPQDFRQVSSVIDVDLLPETHRRVRLHKHGSDRPLGFYIRDGMSVRVAPQGLERVPGIFISRLVRGGLAESTGLLAVSDEILEVNGIEVAGKTLDQVTDMMVANSHNLIVTVKPANQRNNVVRGASGRLTGPPSAGPGPAEPDSDDDSSDLVIENRQPPSSNGLSQGPPCWDLHPGCRHPGTRSSLPSLDDQEQASSGWGSRIRGDGSGFSLSpecifications
NP_001032358.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MARPQRTPARSPDSIVEVKSKFDAEFRRFALPRASVSGFQEFSRLLRAVHQIPGLDVLLGYTDAHGDLLPLTNDDSLHRALASGPPPLRLLVQKREADSSGLAFASNSLQRRKKGLLLRPVAPLRTRPPLLISLPQDFRQVSSVIDVDLLPETHRRVRLHKHGSDRPLGFYIRDGMSVRVAPQGLERVPGIFISRLVRGGLAESTGLLAVSDEILEVNGIEVAGKTLDQVTDMMVANSHNLIVTVKPANQRNNVVRGASGRLTGPPSAGPGPAEPDSDDDSSDLVIENRQPPSSNGLSQGPPCWDLHPGCRHPGTRSSLPSLDDQEQASSGWGSRIRGDGSGFSL | |
RUO | |
PARD6A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
50855 | |
PARD6A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PAR-6A/PAR6/PAR6C/PAR6alpha/TAX40/TIP-40 | |
PARD6A | |
Recombinant | |
wheat germ expression system |