missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PAK1 kinase domain (Q13153, 248 a.a. - 545 a.a.) Partial Recombinant Protein

Codice prodotto. 16111850
Cambia vista
Click to view available options
Quantity:
10 μg
Dimensione della confezione:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
16111850 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16111850 Fornitore Abnova™ N. del fornitore P5382.10ug

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Used for Func, SDS-PAGE

PAK proteins are critical effectors that link RhoGTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, include PAK1, PAK2, PAK3 and PAK4. These proteins serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK1 regulates cell motility and morphology. Alternativelt spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifica

Accession Number Q13153
Concentration 1 mg/ml
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 5058
Molecular Weight (g/mol) 33.7kDa
Name PAK1 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing Loading 8 ug protein in SDS-PAGE
Quantity 10 μg
Immunogen GPHMSDEEILEKLRSIVSVGDPKKKYTRFEKIGQGASGTVYTAMDVATGQEVAIKQMNLQQQPKKELIINEILVMRENKNPNIVNYLDSYLVGDELWVVMEYLAGGSLTDVVTETCMDEGQIAAVCRECLQALEFLHSNQVIHRDIKSDNILLGMDGSVKLTDFGFCAQITPEQSKRSTMVGTPYWMAPEVVTRKAYGPKVDIWSLGIMAIEMIEGEPPYLNENPLRALYLIATNGTPELQNPEKLSAIFRDFLNRCLEMDVEKRGSAKELLQHQFLKIAKPLSSLTPLIAAAKEATKNNH The first 4 residues GPHM are from Turbo3C Protease cleavage site. The underlined T is phosphorylated T423.
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC130000/MGC130001/PAKalpha
Common Name PAK1
Gene Symbol PAK1
Biological Activity Specific activity: 32,638pmoles/min/μg
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Liquid
Vedi altri risultati Mostra meno risultati
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.