Learn More
Abnova™ Human OPHN1 Full-length ORF (NP_002538.1, 1 a.a. - 802 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004983-P01.25ug
Informations supplémentaires : Weight : 0.00010kg
Description
This gene encodes a Rho-GTPase-activating protein that promotes GTP hydrolysis of Rho subfamily members. Rho proteins are important mediators of intracellular signal transduction, which affects cell migration and cell morphogenesis. Mutations in this gene are responsible for OPHN1-related X-linked mental retardation with cerebellar hypoplasia and distinctive facial dysmorhphism. [provided by RefSeq]
Sequence: MGHPPLEFSDCYLDSPDFRERLKCYEQELERTNKFIKDVIKDGNALISAMRNYSSAVQKFSQTLQSFQFDFIGDTLTDDEINIAESFKEFAELLNEVENERMMMVHNASDLLIKPLENFRKEQIGFTKERKKKFEKDGERFYSLLDRHLHLSSKKKESQLQEADLQVDKERHNFFESSLDYVYQIQEVQESKKFNIVEPVLAFLHSLFISNSLTVELTQDFLPYKQQLQLSLQNTRNHFSSTREEMEELKKRMKEAPQTCKLPGQPTIEGYLYTQEKWALGISWVKYYCQYEKETKTLTMTPMEQKPGAKQGPLDLTLKYCVRRKTESIDKRFCFDIETNERPGTITLQALSEANRRLWMEAMDGKEPIYHSPITKQQEMELNEVGFKFVRKCINIIETKGIKTEGLYRTVGSNIQVQKLLNAFFDPKCPGDVDFHNSDWDIKTITSSLKFYLRNLSEPVMTYRLHKELVSAAKSDNLDYRLGAIHSLVYKLPEKNREMLELLIRHLVNVCEHSKENLMTPSNMGVIFGPTLMRAQEDTVAAMMNIKFQNIVVEILIEHFGKIYLGPPEESAAPPVPPPRVTARRHKPITISKRLLRERTVFYTSSLDESEDEIQHQTPNGTITSSIEPPKPPQHPKLPIQRSGETDPGRKSPSRPILDGKLEPCPEVDVGKLVSRLQDGGTKITPKATNGPMPGSGPTKTPSFHIKRPAPRPLAHHKEGDADSFSKVRPPGEKPTIIRPPVRPPDPPCRAATPQKPEPKPDIVAGNAGEITSSVVASRTRFFETASRKTGSSQGRLPGDESSpécification
NP_002538.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
118kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MRX60/OPN1 | |
OPHN1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4983 | |
OPHN1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGHPPLEFSDCYLDSPDFRERLKCYEQELERTNKFIKDVIKDGNALISAMRNYSSAVQKFSQTLQSFQFDFIGDTLTDDEINIAESFKEFAELLNEVENERMMMVHNASDLLIKPLENFRKEQIGFTKERKKKFEKDGERFYSLLDRHLHLSSKKKESQLQEADLQVDKERHNFFESSLDYVYQIQEVQESKKFNIVEPVLAFLHSLFISNSLTVELTQDFLPYKQQLQLSLQNTRNHFSSTREEMEELKKRMKEAPQTCKLPGQPTIEGYLYTQEKWALGISWVKYYCQYEKETKTLTMTPMEQKPGAKQGPLDLTLKYCVRRKTESIDKRFCFDIETNERPGTITLQALSEANRRLWMEAMDGKEPIYHSPITKQQEMELNEVGFKFVRKCINIIETKGIKTEGLYRTVGSNIQVQKLLNAFFDPKCPGDVDFHNSDWDIKTITSSLKFYLRNLSEPVMTYRLHKELVSAAKSDNLDYRLGAIHSLVYKLPEKNREMLELLIRHLVNVCEHSKENLMTPSNMGVIFGPTLMRAQEDTVAAMMNIKFQNIVVEILIEHFGKIYLGPPEESAAPPVPPPRVTARRHKPITISKRLLRERTVFYTSSLDESEDEIQHQTPNGTITSSIEPPKPPQHPKLPIQRSGETDPGRKSPSRPILDGKLEPCPEVDVGKLVSRLQDGGTKITPKATNGPMPGSGPTKTPSFHIKRPAPRPLAHHKEGDADSFSKVRPPGEKPTIIRPPVRPPDPPCRAATPQKPEPKPDIVAGNAGEITSSVVASRTRFFETASRKTGSSQGRLPGDES | |
RUO | |
OPHN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |