missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OPHN1 Full-length ORF (NP_002538.1, 1 a.a. - 802 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_002538.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4983 |
Molecular Weight (g/mol) | 118kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16118591
|
Abnova™
H00004983-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16128591
|
Abnova™
H00004983-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a Rho-GTPase-activating protein that promotes GTP hydrolysis of Rho subfamily members. Rho proteins are important mediators of intracellular signal transduction, which affects cell migration and cell morphogenesis. Mutations in this gene are responsible for OPHN1-related X-linked mental retardation with cerebellar hypoplasia and distinctive facial dysmorhphism. [provided by RefSeq]
Sequence: MGHPPLEFSDCYLDSPDFRERLKCYEQELERTNKFIKDVIKDGNALISAMRNYSSAVQKFSQTLQSFQFDFIGDTLTDDEINIAESFKEFAELLNEVENERMMMVHNASDLLIKPLENFRKEQIGFTKERKKKFEKDGERFYSLLDRHLHLSSKKKESQLQEADLQVDKERHNFFESSLDYVYQIQEVQESKKFNIVEPVLAFLHSLFISNSLTVELTQDFLPYKQQLQLSLQNTRNHFSSTREEMEELKKRMKEAPQTCKLPGQPTIEGYLYTQEKWALGISWVKYYCQYEKETKTLTMTPMEQKPGAKQGPLDLTLKYCVRRKTESIDKRFCFDIETNERPGTITLQALSEANRRLWMEAMDGKEPIYHSPITKQQEMELNEVGFKFVRKCINIIETKGIKTEGLYRTVGSNIQVQKLLNAFFDPKCPGDVDFHNSDWDIKTITSSLKFYLRNLSEPVMTYRLHKELVSAAKSDNLDYRLGAIHSLVYKLPEKNREMLELLIRHLVNVCEHSKENLMTPSNMGVIFGPTLMRAQEDTVAAMMNIKFQNIVVEILIEHFGKIYLGPPEESAAPPVPPPRVTARRHKPITISKRLLRERTVFYTSSLDESEDEIQHQTPNGTITSSIEPPKPPQHPKLPIQRSGETDPGRKSPSRPILDGKLEPCPEVDVGKLVSRLQDGGTKITPKATNGPMPGSGPTKTPSFHIKRPAPRPLAHHKEGDADSFSKVRPPGEKPTIIRPPVRPPDPPCRAATPQKPEPKPDIVAGNAGEITSSVVASRTRFFETASRKTGSSQGRLPGDESSpecifications
NP_002538.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
118kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGHPPLEFSDCYLDSPDFRERLKCYEQELERTNKFIKDVIKDGNALISAMRNYSSAVQKFSQTLQSFQFDFIGDTLTDDEINIAESFKEFAELLNEVENERMMMVHNASDLLIKPLENFRKEQIGFTKERKKKFEKDGERFYSLLDRHLHLSSKKKESQLQEADLQVDKERHNFFESSLDYVYQIQEVQESKKFNIVEPVLAFLHSLFISNSLTVELTQDFLPYKQQLQLSLQNTRNHFSSTREEMEELKKRMKEAPQTCKLPGQPTIEGYLYTQEKWALGISWVKYYCQYEKETKTLTMTPMEQKPGAKQGPLDLTLKYCVRRKTESIDKRFCFDIETNERPGTITLQALSEANRRLWMEAMDGKEPIYHSPITKQQEMELNEVGFKFVRKCINIIETKGIKTEGLYRTVGSNIQVQKLLNAFFDPKCPGDVDFHNSDWDIKTITSSLKFYLRNLSEPVMTYRLHKELVSAAKSDNLDYRLGAIHSLVYKLPEKNREMLELLIRHLVNVCEHSKENLMTPSNMGVIFGPTLMRAQEDTVAAMMNIKFQNIVVEILIEHFGKIYLGPPEESAAPPVPPPRVTARRHKPITISKRLLRERTVFYTSSLDESEDEIQHQTPNGTITSSIEPPKPPQHPKLPIQRSGETDPGRKSPSRPILDGKLEPCPEVDVGKLVSRLQDGGTKITPKATNGPMPGSGPTKTPSFHIKRPAPRPLAHHKEGDADSFSKVRPPGEKPTIIRPPVRPPDPPCRAATPQKPEPKPDIVAGNAGEITSSVVASRTRFFETASRKTGSSQGRLPGDES | |
RUO | |
OPHN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
4983 | |
OPHN1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MRX60/OPN1 | |
OPHN1 | |
Recombinant | |
wheat germ expression system |