Learn More
Abnova™ Human OAZ3 Full-length ORF (AAH73949.1, 1 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051686-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. The ornithine decarboxylase antizymes play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. Antizyme expression is auto-regulated by polyamine-enhanced translational frameshifting. In contrast to antizymes 1 and 2, which are widely expressed throughout the body, the expression of this gene product (antizyme 3) is restricted to testis germ cells, and thus it is a possible candidate for heritable forms of human male infertility. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MTVPWRPGKRRITYKEEEDLTLQPRPASSAPESLVGLQEGKSTEQGNHDQLKELYSAGNLTVLATDPLLHQDPVQLDFHFRLTSQTSAHWHGLLCDRRLFLDIPYQALDQGNRESLTATLEYVEEKTNVDSVFVNFQNDRNDRGALLRAFSYMGFEVVRPDHPALPPLDNVIFMVYPLERDVGHLPSEPPSpecifications
AAH73949.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AZ3/OAZ-t/TISP15 | |
OAZ3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51686 | |
OAZ3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTVPWRPGKRRITYKEEEDLTLQPRPASSAPESLVGLQEGKSTEQGNHDQLKELYSAGNLTVLATDPLLHQDPVQLDFHFRLTSQTSAHWHGLLCDRRLFLDIPYQALDQGNRESLTATLEYVEEKTNVDSVFVNFQNDRNDRGALLRAFSYMGFEVVRPDHPALPPLDNVIFMVYPLERDVGHLPSEPP | |
RUO | |
OAZ3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |