Learn More
Abnova™ Human OAZ3 Full-length ORF (AAH73949.1, 1 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH73949.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51686 |
Molecular Weight (g/mol) | 48.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16190253
|
Abnova™
H00051686-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16100263
|
Abnova™
H00051686-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. The ornithine decarboxylase antizymes play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. Antizyme expression is auto-regulated by polyamine-enhanced translational frameshifting. In contrast to antizymes 1 and 2, which are widely expressed throughout the body, the expression of this gene product (antizyme 3) is restricted to testis germ cells, and thus it is a possible candidate for heritable forms of human male infertility. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MTVPWRPGKRRITYKEEEDLTLQPRPASSAPESLVGLQEGKSTEQGNHDQLKELYSAGNLTVLATDPLLHQDPVQLDFHFRLTSQTSAHWHGLLCDRRLFLDIPYQALDQGNRESLTATLEYVEEKTNVDSVFVNFQNDRNDRGALLRAFSYMGFEVVRPDHPALPPLDNVIFMVYPLERDVGHLPSEPPSpecifications
AAH73949.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MTVPWRPGKRRITYKEEEDLTLQPRPASSAPESLVGLQEGKSTEQGNHDQLKELYSAGNLTVLATDPLLHQDPVQLDFHFRLTSQTSAHWHGLLCDRRLFLDIPYQALDQGNRESLTATLEYVEEKTNVDSVFVNFQNDRNDRGALLRAFSYMGFEVVRPDHPALPPLDNVIFMVYPLERDVGHLPSEPP | |
RUO | |
OAZ3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51686 | |
OAZ3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AZ3/OAZ-t/TISP15 | |
OAZ3 | |
Recombinant | |
wheat germ expression system |