missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OAZ3 Full-length ORF (AAH73949.1, 1 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051686-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100263

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. The ornithine decarboxylase antizymes play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. Antizyme expression is auto-regulated by polyamine-enhanced translational frameshifting. In contrast to antizymes 1 and 2, which are widely expressed throughout the body, the expression of this gene product (antizyme 3) is restricted to testis germ cells, and thus it is a possible candidate for heritable forms of human male infertility. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MTVPWRPGKRRITYKEEEDLTLQPRPASSAPESLVGLQEGKSTEQGNHDQLKELYSAGNLTVLATDPLLHQDPVQLDFHFRLTSQTSAHWHGLLCDRRLFLDIPYQALDQGNRESLTATLEYVEEKTNVDSVFVNFQNDRNDRGALLRAFSYMGFEVVRPDHPALPPLDNVIFMVYPLERDVGHLPSEPP
Specifications

Specifications

AAH73949.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AZ3/OAZ-t/TISP15
OAZ3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
51686
OAZ3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTVPWRPGKRRITYKEEEDLTLQPRPASSAPESLVGLQEGKSTEQGNHDQLKELYSAGNLTVLATDPLLHQDPVQLDFHFRLTSQTSAHWHGLLCDRRLFLDIPYQALDQGNRESLTATLEYVEEKTNVDSVFVNFQNDRNDRGALLRAFSYMGFEVVRPDHPALPPLDNVIFMVYPLERDVGHLPSEPP
RUO
OAZ3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers