missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NUP37 Full-length ORF (NP_076962.2, 1 a.a. - 326 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079023-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194013

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

Bidirectional transport of macromolecules between the cytoplasm and nucleus occurs through nuclear pore complexes (NPCs) embedded in the nuclear envelope. NPCs are composed of subcomplexes, and NUP37 is part of one such subcomplex, Nup107-160 (Loiodice et al., 2004 [PubMed 15146057]).[supplied by OMIM]

Sequence: MKQDASRNAAYTVDCEDYVHVVEFNPFENGDSGNLIAYGGNNYVVIGTCTFQEEEADVEGIQYKTLRTFHHGVRVDGIAWSPETRLDSLPPVIKFCTSAADMKIRLFTSDLQDKNEYKVLEGHTDFINGLVFDPKEGQEIASVSDDHTCRIWNLEGVQTAHFVLHSPGMSVCWHPEETFKLMVAEKNGTIRFYDLLAQQAILSLESEQVPLMSAHWCLKNTFKVGAVAGNDWLIWDITRSSYPQNKRPVHMDRACLFRWSTISENLFATTGYPGKMASQFQIHHLGHPQPILMGSVAVGSGLSWHRTLPLCVIGGDHKLLFWVTEV
Specifications

Specifications

NP_076962.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
63.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ22618/MGC5585/p37
NUP37
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
79023
NUP37 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKQDASRNAAYTVDCEDYVHVVEFNPFENGDSGNLIAYGGNNYVVIGTCTFQEEEADVEGIQYKTLRTFHHGVRVDGIAWSPETRLDSLPPVIKFCTSAADMKIRLFTSDLQDKNEYKVLEGHTDFINGLVFDPKEGQEIASVSDDHTCRIWNLEGVQTAHFVLHSPGMSVCWHPEETFKLMVAEKNGTIRFYDLLAQQAILSLESEQVPLMSAHWCLKNTFKVGAVAGNDWLIWDITRSSYPQNKRPVHMDRACLFRWSTISENLFATTGYPGKMASQFQIHHLGHPQPILMGSVAVGSGLSWHRTLPLCVIGGDHKLLFWVTEV
RUO
NUP37
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers