missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NUP37 Full-length ORF (NP_076962.2, 1 a.a. - 326 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_076962.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79023 |
Molecular Weight (g/mol) | 63.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16104023
|
Abnova™
H00079023-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16194013
|
Abnova™
H00079023-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
Bidirectional transport of macromolecules between the cytoplasm and nucleus occurs through nuclear pore complexes (NPCs) embedded in the nuclear envelope. NPCs are composed of subcomplexes, and NUP37 is part of one such subcomplex, Nup107-160 (Loiodice et al., 2004 [PubMed 15146057]).[supplied by OMIM]
Sequence: MKQDASRNAAYTVDCEDYVHVVEFNPFENGDSGNLIAYGGNNYVVIGTCTFQEEEADVEGIQYKTLRTFHHGVRVDGIAWSPETRLDSLPPVIKFCTSAADMKIRLFTSDLQDKNEYKVLEGHTDFINGLVFDPKEGQEIASVSDDHTCRIWNLEGVQTAHFVLHSPGMSVCWHPEETFKLMVAEKNGTIRFYDLLAQQAILSLESEQVPLMSAHWCLKNTFKVGAVAGNDWLIWDITRSSYPQNKRPVHMDRACLFRWSTISENLFATTGYPGKMASQFQIHHLGHPQPILMGSVAVGSGLSWHRTLPLCVIGGDHKLLFWVTEVSpecifications
NP_076962.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKQDASRNAAYTVDCEDYVHVVEFNPFENGDSGNLIAYGGNNYVVIGTCTFQEEEADVEGIQYKTLRTFHHGVRVDGIAWSPETRLDSLPPVIKFCTSAADMKIRLFTSDLQDKNEYKVLEGHTDFINGLVFDPKEGQEIASVSDDHTCRIWNLEGVQTAHFVLHSPGMSVCWHPEETFKLMVAEKNGTIRFYDLLAQQAILSLESEQVPLMSAHWCLKNTFKVGAVAGNDWLIWDITRSSYPQNKRPVHMDRACLFRWSTISENLFATTGYPGKMASQFQIHHLGHPQPILMGSVAVGSGLSWHRTLPLCVIGGDHKLLFWVTEV | |
RUO | |
NUP37 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
79023 | |
NUP37 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ22618/MGC5585/p37 | |
NUP37 | |
Recombinant | |
wheat germ expression system |