missing translation for 'onlineSavingsMsg'
Learn More

Human NT-4 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18039791
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18039791 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18039791 Supplier R&D Systems Supplier No. AF268NA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

NT-4 Polyclonal specifically detects NT-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NT-4
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.4-2 ug/mL
Gene Accession No. P34130
Gene Alias GLC10, GLC1O, neurotrophic factor 4, neurotrophic factor 5, neurotrophin 4, neurotrophin 5 (neurotrophin 4/5), neurotrophin-4, neurotrophin-5, Neutrophic factor 4, NT4, NT-4, NT-4/5, NT-5, NTF4, NTF5, NTF5NT5
Gene Symbols NTF4
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human NT-4 Gly81-Ala210 Accession # P34130
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4909
Test Specificity Detects human NT-4 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) NT-3 and rhBDNF is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.