missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NRG1-beta 1/HRG1-beta 1 Extracellular Domain Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF377-SP
This item is not returnable.
View return policy
Description
Neuregulin-1 beta 1/NRG1 beta 1 Polyclonal specifically detects Neuregulin-1 beta 1/NRG1 beta 1 in Human samples. It is validated for Western Blot.
Specifications
| Neuregulin-1 beta 1/NRG1 beta 1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARIA, GGF, GGF2, Heregulin-1, HGL, HRG, HRG1, HRG1-alpha, HRG1-beta 1, HRGA, MST131, MSTP131, NDF, neuregulin 1, Neuregulin1, Neuregulin1 beta 1, NRG1, NRG1-IT2, SMDF | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3084 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| AAA58639 | |
| NRG1 | |
| E. coli-derived recombinant human NRG1-beta 1/HRG1-beta 1 Ser2-Lys246 Accession # AAA58639 | |
| 25 μg | |
| Primary | |
| Detects human NRG1-beta 1/HRG1-beta 1 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with the EGF domain of recombinant human (rh) NRG1-beta 1/HRG1-beta 1 is observed and less than 1% cross-reactivity with rhNRG-2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction