missing translation for 'onlineSavingsMsg'
Learn More

Human Niemann-Pick Type C2/NPC2 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18722219
Change view
Click to view available options
Quantity:
25 μL
Unit Size:
25µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18722219 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18722219 Supplier R&D Systems Supplier No. AF8644SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Niemann-Pick type C2 Polyclonal specifically detects Niemann-Pick type C2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Niemann-Pick type C2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunohistochemistry 1:50 dilution, Immunocytochemistry 1:150 dilution
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. P61916
Gene Alias EDDM1, Epididymal Protein 1, epididymal secretory protein E1, HE1, HE1epididymal protein 1, Human epididymis-specific protein 1, MGC1333, Niemann-Pick disease type C2 protein, Niemann-Pick disease, type C2, NiemannPick Type C2, NPC2, NP-C2, tissue-specific secretory protein
Gene Symbols NPC2
Host Species Rabbit
Immunogen S. frugiperda insect ovarian cell line Sf21-derived recombinant human Niemann-Pick Type C2/NPC2 Glu20-Leu151 Accession # P61916
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10577
Test Specificity Detects human Niemann-Pick Type C2/NPC2 in direct ELISAs and Western blots.
Target Species Human
Content And Storage 12 months from date of receipt, -20 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 degreesC under sterile conditions after opening.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.